Cyclopentanethiol 98%

Warning

AC43569-0050 AC43569-0250
103533-480EA 95.3 USD
103533-480 103533-478
Cyclopentanethiol 98%
Cyclopentanethiol
Formula: C₅H₁₀S
Boiling Pt: 129…131 °C (760 mmHg)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001369
CAS Number: 1679-07-8
UN: 3336
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
NMR (31P-NMR) ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR