Iodocyclopentane 98% stabilized

Warning

Synonyms: Cyclopentyl iodide

Stabilization: Stabilised with copper

AC38173-0050 AC38173-0250
103532-790EA 171 USD
103532-790 103532-788
Iodocyclopentane 98% stabilized
Iodocyclopentane
Formula: C₅H₉I
MW: 196.03 g/mol
Boiling Pt: 166…167 °C (760 mmHg)
Density: 1.695 g/cm³
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039446
CAS Number: 1556-18-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR