2-Quinolinecarbonitrile 97%

Warning

Synonyms: Quinoline-2-carbonitrile

AC44958-0050 AC44958-0010
103533-982EA 82.7 USD
103533-982 103533-984
2-Quinolinecarbonitrile 97%
2-Quinolinecarbonitrile
Formula: C₁₀H₆N₂
MW: 154.17 g/mol
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00134341
CAS Number: 1436-43-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 92 to 96 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR