m-Ethyltoluene 98%

Danger

Synonyms: 3-Ethyltoluene1-Ethyl-3-methylbenzene

AC11900-0100 AC11900-0250
103530-648EA 88.6 USD
103530-648 103530-650
m-Ethyltoluene 98%
m-Ethyltoluene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 158…159 °C (760 mmHg)
Melting Pt: 97…95 °C
Density: 0.86 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00009259
CAS Number: 620-14-4
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Loss on drying ≤5 %
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR