α,α'-Dibromo-m-xylene 97%

Danger

Synonyms: 1,3-Bis(bromomethyl)benzenem-Xylylene dibromide

AC15505-0050 AC15505-0250
103531-406EA 70.8 USD
103531-406 103531-408
α,α'-Dibromo-m-xylene 97%
α,α'-Dibromo-m-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 135…140 °C (20 mmHg)
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000178
CAS Number: 626-15-3
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR