Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Phenylbenzyl bromide ≥90%

Supplier: ACROS ORGANICS MS

Danger
Synonyms: Benzhydryl bromideα-Phenylbenzyl bromideBromodiphenylmethane

AC37346-1000 AC37346-5000
103532-686EA 28.84 USD InStock
103532-686 103532-688
α-Phenylbenzyl bromide ≥90%
α-Phenylbenzyl bromide


Formula: (C₆H₅)₂CHBr
MW: 247,13 g/mol
Boiling Pt: 184°C (20 mm Hg)
Melting Pt: 33…42 °C
Density: 1.491 g/cm³ (13 °C)
Flash Pt: >110°C
Storage Temperature: Ambient
MDL Number: MFCD00000134
CAS Number: 776-74-9
EINECS: 212-279-8
UN: 1770
ADR: 8,II
Order Now
Specification Test Results
Appearance Orange to brown Low melting crystalline mass or solid
Infrared spectrum Conforms
Melting point 33°C to 42°C
GC ≥88 %


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty