3-Fluoroanisole 98%

Warning

Synonyms: 1-Fluoro-3-methoxybenzene

AC17189-1000 AC17189-0050 AC17189-0250
103531-666EA 129.2 USD
103531-666 103531-662 103531-664
3-Fluoroanisole 98%
3-Fluoroanisole
Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 158 °C (743 mmHg)
Melting Pt: –35 °C
Density: 1.104 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000335
CAS Number: 456-49-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR