3,4-Dichlorothiophenol 97%

Warning

Synonyms: 3,4-Dichlorobenzene-1-thiol3,4-Dichlorobenzenethiol1,2-Dichloro-4-mercaptobenzene1,2-Dichloro-4-benzenethiol

AC16285-0050 AC16285-0010
103531-516EA 16.7 USD
103531-516 103531-518
3,4-Dichlorothiophenol 97%
3,4-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 119…120 °C (10 mmHg)
Melting Pt: 87…89 °C
Density: 1.4 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004840
CAS Number: 5858-17-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR