3,4-Dichlorobenzonitrile 97%

Warning

AC43499-0050
103533-456EA 57.9 USD
103533-456
3,4-Dichlorobenzonitrile 97%
3,4-Dichlorobenzonitrile
Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00016379
CAS Number: 6574-99-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 69.0 to 74.0 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR