2-Amino-5-hydroxybenzoic acid 99%

Warning

AC31031-0010 AC31031-0100
103532-300EA 22.3 USD
103532-300 103532-302
2-Amino-5-hydroxybenzoic acid 99%
2-Amino-5-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 233…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00007870
CAS Number: 394-31-0
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR