1-Bromo-2,4,5-trifluorobenzene 98%

Warning

Synonyms: 2,4,5-Trifluorobromobenzene

AC27116-0250 AC27116-0050
103532-058EA 57.18 USD
103532-058 103532-060
1-Bromo-2,4,5-trifluorobenzene 98%
1-Bromo-2,4,5-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 143…144 °C (760 mmHg)
Melting Pt: –19 °C
Density: 1.802 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00000306
CAS Number: 327-52-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR