3,3-Dimethyl-1-butene 95%

Danger

AC15272-2500 AC15272-0500
103531-372EA 29.9 USD
103531-372 103531-374
3,3-Dimethyl-1-butene 95%
3,3-Dimethyl-1-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 41 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.65 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008853
CAS Number: 558-37-2
EINECS: 209-195-9
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR