Neopentylamine 97%

Danger

Synonyms: 1-Amino-2,2-dimethylpropane

AC17369-0250 AC17369-1000
103531-680EA 138 USD
103531-680 103531-682
Neopentylamine 97%
Neopentylamine
Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 79…82 °C (760 mmHg)
Density: 0.74 g/cm³
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008134
CAS Number: 5813-64-9
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR