4,4-Dimethyl-2-pentanone 99%

Danger

Synonyms: 4,4-Dimethylpentan-2-one

AC15971-0050 AC15971-0250 AC15971-1000
103530-030EA 430 USD
103530-030 103530-032 103530-028
4,4-Dimethyl-2-pentanone 99%
4,4-Dimethyl-2-pentanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 125…130 °C (760 mmHg)
Melting Pt: –64 °C
Density: 0.804 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008854
CAS Number: 590-50-1
EINECS: 209-685-2
UN: 1224
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR