3-Methyl-2-pentanone 99%

Danger

AC12738-0050 AC12738-0010 AC12738-0250
103530-822EA 194 USD
103530-822 103530-818 103530-820
3-Methyl-2-pentanone 99%
3-Methyl-2-pentanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 118 °C (758 mmHg)
Density: 0.81 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009336
CAS Number: 565-61-7
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR