Osmium 99.9% (trace metals basis)

Danger

AC38516-0050 AC38516-0010
103532-860EA 772.15 USD
103532-860 103532-858
Osmium 99.9% (trace metals basis)
Osmium
Formula: Os
Boiling Pt: 5027 °C (760 mmHg)
Melting Pt: 3045 °C
Storage Temperature: Ambient
MDL Number: MFCD00011147
CAS Number: 7440-04-2
EINECS: 231-114-0
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Blue-grey Powder
Particle Size/Sieve analysis ≤100 mesh
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR