Streptavidin, AP (Alkaline Phosphatase), Jackson ImmunoResearch
Supplier: Jackson Immunoresearch Laboratories
016-050-084
102643-972EA 248.62 USD
102643-972
Streptavidin, AP (Alkaline Phosphatase), Jackson ImmunoResearch
Proteins and Peptides
Steptavidin is a bacterial protein isolated from Streptomyces avidinii, is similar to egg-white avidin in its ability to bind biotin and used as a replacement for egg-white avidin because of its more favorable chemical properties.
Caution: For In vitro research use only. It is not a medical device and it is not intended for diagnostic or therapeutic purposes.