Pgp-Glo Assay System
Supplier: Promega
V3591
PAV3591EA 417.69 USD
PAV3591
Pgp-Glo Assay System
The Pgp-Glo Assay Systems provide the necessary reagents for performing luminescent P-glycoprotein (Pgp) ATPase assays.
  • Complete Set of Reagents for Performing Luminescent P-glyoprotein (Pgp) ATPase Assays
  • Simple, fast protocol
  • "Glow-type" signal allows processing of multiple samples
  • Assay available with, or without, recombinant Pgp
The Pgp-Glo Assay Systems provide the necessary reagents for performing luminescent P-glycoprotein (Pgp) ATPase assays. Pgp, also known as MDR1 and ABCB1, is a 170kDa integral plasma membrane protein that functions as an ATP-dependent drug efflux pump and plays an important role in multidrug resistance and certain adverse drug-drug interactions. Compounds that interact with Pgp can be identified as stimulators or inhibitors of its ATPase activity. Compounds that are substrates for transport by Pgp typically stimulate its ATPase activity. The Pgp-Glo Assay detects the effects of compounds on recombinant human Pgp in a cell membrane fraction. The assay relies on the ATP dependence of the light-generating reaction of firefly luciferase. ATP is first incubated with Pgp; then the Pgp ATPase reaction is stopped, and the remaining unmetabolized ATP is detected as a luciferase-generated luminescent signal. Pgp-dependent decreases in luminescence reflect ATP consumption by Pgp; thus the greater the decrease in signal, the higher the Pgp activity. Accordingly, samples containing compounds that stimulate the Pgp ATPase will have significantly lower signals than untreated samples.