IL-1β (human), ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-130A
76199-134EA 741.09 USD
76199-134
IL-1β (human), ELISA kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of IL-1β in culture supernatant, plasma, serum, and urine of human origin.
  • Sensitive - quantify down to 1 pg/mL
  • Efficient - only 50 µl diluted sample required saving precious materials
  • Specific - no cross-reactivity with related cytokines
  • Results from up to 40 samples in duplicate in just 3 hours
  • Ready-to-use reagents and pre-coated plates reduce errors and save your time
The IL-1β (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in less than 3 hours. Absorbance is read at 450 nm. Quantify 1 pg/mL IL-1β with lot sample volume needed saving you precious sample.
Ordering information: Contains Standard, Standard Diluent Buffer, Coated 96-well plate, Conjugate, Streptavidin-HRP, Streptavidin-HRP Diluent, Wash Buffer Concentrate, TMB Solution, Stop Solution, Plate Sealers.