Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureYield Plasmid Miniprep Start-Up Kit, 110V Electrical (750 preps, manifold and free vacuum pump), 1 each, Promega®

Supplier: Promega Corporation
A6810
PureYield Plasmid Minipreps rapidly isolate up to 15ug of transfection-quality plasmid DNA from 600ul to 3ml cultures. Start-up kit contains 750 minipreps, vacuum manifold and free vacuum pump.

  • Plasmid Miniprep System, Vacuum Manifold and Free Vacuum Pump
  • Up to 15µg plasmid DNA from 10ml cultures
  • Transfection-quality plasmid DNA in 10 minutes
  • No tedious, gravity-drip columns or post-elution alcohol precipitation

The PureYield Plasmid Miniprep System is designed to rapidly isolate highly pure plasmid DNA. The system provides a rapid method for purification of up to 15ug of plasmid DNA from 600ul to 3ml of bacteria culture. Plasmid DNA can be purified in as little as 10 minutes. The PureYield Plasmid Miniprep System incorporates a unique Endotoxin Removal Wash designed to remove substantial amounts of protein, RNA and endotoxin contaminants from purified plasmid DNA. Removal of contaminants improves the robustness of sensitive applications such as eukaryotic transfection, in vitro transcription and coupled in vitro transcription/translation. Purification is achieved without isopropanol precipitation of purified plasmid DNA or extensive centrifugation, providing rapid purification from a single method. The system has been designed for use with centrifugation or vacuum (e.g., the Vac-Man Laboratory Vacuum Manifold). The Start-Up Kit provides 750 minipreps, a vacuum manifold and a free vacuum pump.




 

ORDER

Expand All / Collapse All
Description Size Includes Supplier No. VWR Catalog Number Unit Price Quantity
PureYield Plasmid Miniprep Start-Up Kit, 110V Electrical (750 preps, manifold and free vacuum pump), 1 each 1 each Manifold and Free Vacuum Pump, 110V A6810 PAA6810 Case of 750 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Storage Temperature 15°C to 30°C
Storage Conditions Store all system components at 22 to 25°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty