Actinomycin D ≥93%

AC29494-1000 AC29494-0010
103532-130EA 1128 USD
103532-130 103532-128
Actinomycin D ≥93%
Actinomycin D

Red Crystalline powder

Formula: C₆₂H₈₆N₁₂O₁₆
MW: 1255.5 g/mol
Melting Pt: 245…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005033
CAS Number: 50-76-0
UN: 3462
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR