Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

Cork Borers

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (470121-860)

Supplier:  VWR International
Description:   This sharpened, nickel-plated borer has attached handles that are numbered for easy identification.
Supplier:  American Scientific
Description:   Nickel-plated brass handles and borer tubes punch a variety of hole sizes.
Small Business Enterprise
Catalog Number: (52924-950)

Supplier:  Humboldt Manufacturing
Description:   The handy tool is used to sharpen borers from 3/16" to 15/16" (5 mm to 24 mm)
Catalog Number: (23457-112)

Supplier:  Boekel
Description:   Made of nickel-plated steel for a sharper, long-lasting cutting edge
Supplier:  Humboldt Manufacturing
Description:   These cork borer sets are used to bore holes in corks and rubber stoppers.
Small Business Enterprise
Catalog Number: (23457-060)

Supplier:  Boekel
Description:   Made of brass tubing with strong permanent handles
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 6  of 6
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping