DBI-SALA Nano-Lok™ Extended Length Self Retracting Web Lifelines with Anchor Hooks, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3101583
76111-448EA 364.51 USD
76111-448
DBI-SALA Nano-Lok™ Extended Length Self Retracting Web Lifelines with Anchor Hooks, ORS Nasco
Fall Protection Fall Arresters
The lightest, most compact unit in its class is easy to use, relocate, store, and can be easily used as a lanyard replacement.
  • Swiveling anchorage loop with aluminum carabiner limits lifeline twisting for smoother operation
  • Nano-Lok™ is ergonomically designed for ease-of-use and is ideal for direct connection to most harnesses
  • Automatically locks and closes for added safety, and its aluminum construction reduces overall weight to add comfort
  • Compact and durable housing is designed to withstand the demands of challenging jobsites
  • Quick activating speed sensing brake system stops a falling worker within inches, providing more protection at low heights
  • Built-in RFID identification tag helps the user record and store information on inspections
  • Wide variety of configurations available
The synthetic lifeline reduces overall weight and the retractable design provides freedom of movement, keeps the lifeline out of the worker's way, and reduces dragging, snagging, and trip falls. Specialized built-in external absorption system maintains safe fall arrest forces for users and tools up to 420 lbs., and provides a visual alert that the SRL has been involved in a fall.