DBI-SALA® Rebel™ Self Retracting Cable Lifelines, Capital Safety
Supplier: DBI/Sala
098-3590550 098-3590016 098-3590500 098-3590590
76111-506EA 871 USD
76111-506 76111-502 76111-504 76111-510
DBI-SALA® Rebel™ Self Retracting Cable Lifelines, Capital Safety
Fall Protection Fall Lanyards and Ropes
  • Abrasion-resistant and extremely durable lifeline for added field longevity
  • Retractable design provides freedom of movement and reduces dragging, snagging, and trip falls
  • Rugged, thermoplastic housing is compact and lightweight
  • Side lifeline exit with ergonomic handle enables smooth operation and reduces cable wear
Swiveling self-locking snap hook with impact indicator automatically locks and closes for added safety, swivels to limit lifeline twisting, and provides visual indication of fall arrest. Speed sensing, all-metal brake system stops a falling worker within inches and provides smooth consistent operation, allowing the user to maintain normal work pace.
Certifications: Meets all applicable industry standards including OSHA and ANSI Z359.1.