DBI-SALA® ExoFit NEX™ Climbing Harnesses, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-1113034 098-1113040 098-1113031
76111-282EA 581.62 USD
76111-282 76111-284 76111-286
DBI-SALA® ExoFit NEX™ Climbing Harnesses, ORS Nasco
Fall Protection Fall Harnesses
  • Aluminum hardware
  • ExoFit NEX™ vest style harness, highest level of safety and the utmost comfort
  • Spring loaded torso buckles for quick and easy adjustment
Certifications: Meets or exceeds all applicable industry standards, including OSHA and ANSI. Also including the stringent ANSI Z359.