Anti-Slc9a3r1 Rabbit Polyclonal Antibody

Supplier: BioVision
3521-30T
76208-950EA 103.88 USD
76208-950
Anti-Slc9a3r1 Rabbit Polyclonal Antibody
NHE3 regulatory factor 1 (NHERF-1) is a multifunctional adaptor protein that interacts with NHE3 via its PDZ domains. The amino-terminal PDZ domain regulates the dimerization of NHERF-1 in vivo. G protein-coupled receptor kinase 6A phosphorylates NHERF-1 at Ser 289 via a PDZ domain-mediated interaction.

Type: Primary
Antigen: Slc9a3r1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR