Anti-CXCR5 Rabbit Polyclonal Antibody

Supplier: BioVision
3419-30T
76209-060EA 103.91 USD
76209-060
Anti-CXCR5 Rabbit Polyclonal Antibody
Chemokines play an important role in inflammation and critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines. CXCR5 is one of the CXCR receptors, which is expressed in a variety of cells.

Type: Primary
Antigen: CXCR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR