Anti-EPCAM Rabbit Monoclonal Antibody [clone: EGP40/1555R]

Supplier: Enzo Life Sciences
ABS364-0100
76201-816EA 589.97 USD
76201-816
Anti-EPCAM Rabbit Monoclonal Antibody [clone: EGP40/1555R]
Antibodies
Recombinant monoclonal antibody

  • Recommended Applications: Flow Cytometry, IF, IHC (PS), WB
  • Species reactivity: Human

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: EGP40/1555R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR