Anti-HSP90AA1 Rabbit Polyclonal Antibody

Supplier: BioVision
3389-30T
76208-772EA 103.91 USD
76208-772
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Heat shock proteins (HSPs) are ubiquitously expressed in all organisms. A major function of HSP90 and other HSPs is to act as molecular chaperones. HSP90 forms a complex with glucocorticoid receptor (GR), rendering the non ligand-bound receptor transcriptionally inactive. HSP 90 binds the GR as a heterocomplex composed of either HSP56 or cyclophilin-40, forming an aporeceptor complex. HSP90 also exists as a dimer with other proteins such as p60/sti1 and p23, forming an aporeceptor complex with estrogen and androgen receptors.

Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR