Anti-CAPN2 Rabbit Polyclonal Antibody

Supplier: BioVision
3372-30T
76209-050EA 103.91 USD
76209-050
Anti-CAPN2 Rabbit Polyclonal Antibody
Calpains are cytosolic, calcium-dependent, cysteine proteases that are ubiquitously expressed in mammalian and avian tissues. Calpain-1 and 2 consist of a common small subunit and a large variable subunit. Domains in the large subunit include the amino terminal domain I, the proteinase domain II, domain III, and the EF-hand domain IV. Type I calpain requires micromolar amounts of calcium, while type II calpain requires millimolar amounts of calcium.

Type: Primary
Antigen: CAPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR