Anti-TP53INP1 Rabbit Polyclonal Antibody

Supplier: BioVision
3340-30T
76208-770EA 103.91 USD
76208-770
Anti-TP53INP1 Rabbit Polyclonal Antibody
p53DINP1 (p53-dependent damage-inducible nuclear protein 1) encodes two proteins termed p53DINP1-α (27 kDa) and p53DINP1-β (18 kDa). p53DINP1 may regulate p53-dependent apoptosis throμgh phosphorylation at Ser46 and induction of p53AIP1. p53DINP1 is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.

Type: Primary
Antigen: TP53INP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR