Anti-GSAP Rabbit Polyclonal Antibody

Supplier: BioVision
3308-30T
76209-010EA 103.91 USD
76209-010
Anti-GSAP Rabbit Polyclonal Antibody
GSAP, gamma-secretase activating protein interacts with gamma secretase which chops up the amyloid precursor protein to senile plaques in Alzheimer’s disease. GSAP was found to increase the production amyloid peptide. Immunohistochemistry to identify GSAP in brain can demonstrate its abundance in certain regions of damage, and is usually morphologically changed throughout the progression of Alzheimer's disease. GSAP can be used as a qualitative biomarker of Alzheimer's disease progression, and is regularly considered one of the best targets of therapies to diminish the effects of Alzheimer's disease in affected individuals.

Type: Primary
Antigen: GSAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR