Anti-LMNA protein Rabbit Polyclonal Antibody

Supplier: BioVision
3267-30T
76209-354EA 103.91 USD
76209-354
Anti-LMNA protein Rabbit Polyclonal Antibody
Lamins are nuclear membrane structural components that are important in maintaining normal cell functions such as cell cycle control, DNA replication and chromatin organization. Lamin A is specifically cleaved by caspase-6 and therefore serves as a marker for caspase-6 activation. During apoptosis the 70 kDa lamin A is cleaved to a large (40-45 kDa) and small (23 kDa) fragment. The cleavage of lamins results in nuclear disregulation and cell death.

Type: Primary
Antigen: LMNA protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR