Anti-Tctn2 Rabbit Polyclonal Antibody

Supplier: BioVision
3262-30T
76209-350EA 103.91 USD
76209-350
Anti-Tctn2 Rabbit Polyclonal Antibody
TCTN2 is a member of the Tectonic protein family. Tectonic proteins are transmembrane proteins that regulate the Hedgehog (Hh)-signaling pathway. Defects in TCTN2 can lead to Meckel syndrome type 8. This disorder is characterized by a combination of renal cysts and variable associated features which includes developmental anomalies of the central nervous system, hepatic ductal dysplasia and cysts, and polydactyly.

Type: Primary
Antigen: Tctn2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR