Anti-GNAI2 Rabbit Polyclonal Antibody

Supplier: BioVision
3249-30T
76209-576EA 103.91 USD
76209-576
Anti-GNAI2 Rabbit Polyclonal Antibody
Gαi2 is a 40-41 kDa alpha-subunit of heterotrimeric protein that shares sequence homology with Gαi3 and Gαi1. All these G proteins can be ADP-ribosylated at the carboxy terminal Cys of the CAAX sequence, interfering with receptor coupling. Gαi2 is expressed widely in tissues and cell lines. Activation of PLC-β by Gαi-coupled receptors is the result of release of Gβg subunits from Gαi. Gαi2 regulates potassium channel activation. Mutation at Arg179 has been identified in tumors. This and the Q205L mutant inhibit adenylyl cyclase.

Type: Primary
Antigen: GNAI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR