Anti-SIRT3 Rabbit Polyclonal Antibody

Supplier: BioVision
3223-30T
76209-150EA 103.91 USD
76209-150
Anti-SIRT3 Rabbit Polyclonal Antibody
Silent information regulator (Sir2)-like family deacetylases (also known as sirtuins) are highly conserved proteins and have important roles in the regulation of metabolism, inflammation, cellular survival growth and differentiation. SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Yeast sirtuin proteins are found to regulate epigenetic gene silencing. Sirtuins are NAD-dependent protein ADP-ribosyl transferase which catalyzes the transfer of ADP-ribosyl groups onto target proteins.

Type: Primary
Antigen: SIRT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR