Anti-GFAP Rabbit Polyclonal Antibody

Supplier: BioVision
3206-30T
76209-148EA 103.91 USD
76209-148
Anti-GFAP Rabbit Polyclonal Antibody
GFAP, Glial fibillary acidic protein is an intermediate filament protein. It was found in astrocytes cells as a cell specific marker in the central nervous system development. GFAP is defective in Alexander disease. But it is highly expressed in Astrogliosis which is a result of some diseases, such as AIDS, dementia and inflammatory demyelination diseases.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR