Anti-AIFM1 Rabbit Polyclonal Antibody

Supplier: BioVision
3202-30T
76209-144EA 103.91 USD
76209-144
Anti-AIFM1 Rabbit Polyclonal Antibody
AIF (apoptosis inducing factor) was recently identified that causes chromatin condensation and DNA fragmentation. AIF translocates to the nucleus when apoptosis is induced and induces mitochondria to release the apoptogenic proteins cytochrome c and caspase-9. AIF induces chromatin condensation and DNA fragmentation, which are the hallmarks of apoptosis. AIF is highly conserved between human and mouse and widely expressed.

Type: Primary
Antigen: AIFM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR