Anti-Capn1 Rabbit Polyclonal Antibody

Supplier: BioVision
3189-30T
76209-136EA 103.91 USD
76209-136
Anti-Capn1 Rabbit Polyclonal Antibody
Calpains are cytosolic, calcium-dependent, cysteine proteases that are ubiquitously expressed in mammalian and avian tissues. Type I calpain requires micromolar amounts of calcium, while type II calpain requires millimolar amounts of calcium.

Type: Primary
Antigen: Capn1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat;Bovine;Pig;Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR