- Home
- Tubing
- Transfer Tubing
- GloMax 96 Tubing Replacement Kit for Injectors, Promega
GloMax 96 Tubing Replacement Kit for Injectors, Promega
Supplier: Promega
E4822
PAE4822EA
766.31
USD
PAE4822
GloMax 96 Tubing Replacement Kit for Injectors, Promega
Tubing
Transfer Tubing
GloMax 96 Microplate Luminometer represents instrumentation and software in a complete solution that includes bioluminescent assays, protocols and support.
- Instrument Accessories
- Instrument is discontinued
- Accessories available until November 1, 2021, pending part availability
The GloMax 96 Microplate Luminometer represents instrumentation and software in a complete solution that includes bioluminescent assays, protocols and support. The GloMax 96 is a state-of-the-art microplate luminometer that meets the needs for high sensitivity and broad dynamic range for all luminescence applications. Available with up to two reagent injectors, the GloMax 96 Microplate Luminometer is a versatile system designed to perform both flash- and glow-type luminescence assays. The GloMax 96 Microplate Luminometer also includes a power cable, data cable, Quick Protocol card, 5 white 96-well microplates, and software required to operate the instrument. This instrument requires the use of a computer with Microsoft Excel. The GloMax 96 Microplate Luminometer provides superior sensitivity and precision for all luminescent assays. Proprietary circuitry and an advanced photon-counting photomultiplier tube (PMT) provide unmatched signal-to-noise ratios. The option of an intelligently designed internal auto-injection system is an added convenience. Connections, priming and flushing are greatly simplified because up to two reagent injectors are designed to fit next to the plate detection module. This arrangement minimizes dispensing problems, simplifies maintenance and reduces service calls. The dispensing design also includes features that help the user save valuable time and reagents, including an open architecture that enables the user to inspect all tubing and tips during operation. The software features preloaded protocols to run Promega assays. Setup wizards guide the user through a brief process when establishing new protocols. New users can set up protocols and operate the instrument without a steep learning curve. The user can quickly select the protocol of interest and begin running assays with a minimum of modification. Direct-to-Excel-based software reports data directly to an Excel spreadsheet, where data can be analyzed quickly and easily. An Excel macro assists in data analysis for Dual-Luciferase assays. The GloMax 96 Microplate Luminometer Light Plate provides a quick and easy means to verify the performance of the GloMax 96 Microplate Luminometer. Users can check the sensitivity, reproducibility and linearity. The Light Plate consists of three highly stable light sources that simulate luminescent samples at signal levels spanning four decades. The unit is powered by a battery that is widely available and easy to replace. The GloMax 96 Tubing Replacement Kit for Injectors contains parts for replacement of two complete fluid paths in the GloMax 96 Microplate Luminometers configured with reagent injectors. Items contained within the kit include two sets of tubing and a pack of 10 injector tips. To learn more about the GloMax Systems, please visit: www.promega.com/products/instruments/.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart