    Sort by:
  • Search Within Results
Cystein Derivatised Silica Gel
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  SILICYCLE INC.
Description:   SiliaMetS® Cysteine is the silica bound equivalent of the amino acid Cysteine. By attaching the molecule to the backbone via the amino group, the thio...