Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crazy Circuits With Squishy Circuits™

Supplier: BROWN DOG GADGETS
K1025
Combine the power of Arduino with Squishy Circuits™.

  • Create your own programs or use one of the many online guides or tutorials
  • Make your own dough based lighting and sound system or even your own dough-based game controller!
  • Crazy Circuits parts are compatible with LEGO™ and similar brick building systems
  • Crazy Circuits parts come 100% ready to go out of the box, which means no soldering or preparation required

The included Touch Board provides users with the ability to turn the included Squishy Circuits™ Dough into interactive programing projects. This kit covers topics such as circuitry, programming, logic, design, sewable technology, art, and design. Detailed directions, project guides, educational resources, and videos can be found in our online database.

Ordering information: Includes Crazy Circuits Chips; Touch Board and USB Cable; Five Jumbo 10mm LEDs (Red, Amber, Green, Blue, White); Piezoelectric Speaker; Squishy Circuits™ Conductive Dough - 6 Pack; Conductive Nylon Tape - 1/8", 16 feet; Five Alligator Clips; LEGO™ Baseplate (8 x 16 studs); and Plastic Organizer.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Crazy Circuits With Squishy Circuits™ K1025 470234-686 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty