Heparin, Biotin Conjugate (from porcine intestinal mucosa), solid, Calbiochem®
Supplier: MilliporeSigma
375054-10MG
80502-706EA 252.54 USD
80502-706
Heparin, Biotin Conjugate (from porcine intestinal mucosa), solid, Calbiochem®
Heparin, Biotin Conjugate
Biotinylated heparin containing 5 – 8 mole-percent biotin.
Biotin attachment is through a 12-atom spacer arm containing a cleavable disulfide group, molecular weight of 15000.