Would you like to log in again?
pVL1392, pVL1393 Transfer Vectors, Expression Systems
Supplier: Expression Systems
91-030 91-013 91-012
pVL1392 and pVL1393 are transfer vectors designed for high-level expression of a cloned gene driven by the strong Autographa californica multiple nucleopolyhedrovirus (AcMNPV) polyhedrin promoter (pPolh).
These are pUC8 based plasmids containing the AcMNPV polyhedrin locus lacking the majority of the polyhedrin open reading frame. A multiple cloning site (MCS) sequence has been inserted 37 nucleotides downstream from the original polyhedrin ATG start codon, which has been mutated to ATT.
The vectors share identical sequences, with the exception that they each have the same MCS in opposing orientations. A start and stop codon must be provided with the gene that is to be cloned into the MCS. For maximal recombinant protein expression, the distance between the cloning site and the ATG start of the inserted gene should be kept to a minimum, and should not exceed 100 nucleotides.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
pVL1392 Transfer Vector 91-012 103194-176 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
pVL1393 Transfer Vector 91-013 103194-178 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
pVL1392, pVL1393 Transfer Vector Set 91-030 103194-180 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty