Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Metro® Mobile Space Saver Worktables, Stainless Steel

Supplier: Metro
MWTS2448FS MWTS2448US MWTS2460US MWTS2460FS MWTS2436US MWTS2436FS
Maximize every inch with Metro® Mobile Space Saver Worktables.

  • Durable stainless steel components
  • 5" polymer corrosion resistant
  • Available with a solid bottom shelf or 3-sided frame
  • Easy assembly

Made from durable stainless steel components to stand up to the harshest applications and environments, sturdy construction provides a robust 800 lbs. load rating.

The 1" diameter posts and patented Super Erecta Shelf construction assure a rigid, sway-free table. Heavy-duty 14-gauge stainless steel virtually eliminates any "drumming."

Easily assembles in minutes without the use of special tools.



 

ORDER

Expand All / Collapse All

Solid Shelf
Length Width Supplier No. VWR Catalog Number Unit Price Quantity
36" 24" MWTS2436FS 76181-286 Each Retrieving
48" 24" MWTS2448FS 76181-288 Each Retrieving
60" 24" MWTS2460FS 76181-290 Each Retrieving

3-Sided Frame
Length Width Supplier No. VWR Catalog Number Unit Price Quantity
36" 24" MWTS2436US 76181-330 Each Retrieving
48" 24" MWTS2448US 76181-332 Each Retrieving
60" 24" MWTS2460US 76181-334 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty