Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PIPETMAN® F Fixed Volume Single Channel Pipettor, Gilson®

Supplier: GILSON, INC.
PIPETMAN® F
F123787 F123788 F123775 F123604 F123605 F123778 F123789 F123790 F123772 F123784 F123770 F123771 F123606 F123607
Fixed volume air-displacement pipettor PIPETMAN® F.

  • Long product life and high resistance to aggressive chemicals thanks to stainless steel and PVDF components
  • Ultimate security and consistent results with fixed volume setting
  • GLP compliance: No risk of volume selection error
  • For clinical diagnostics, quality control, and any routine testing

PIPETMAN® F is a fixed-volume, air-displacement pipettor that eliminates the risk of volume selection errors. Thirteen robust models cover a large volume range, from 2 µL to 1000 µL, all with Gilson's legendary accuracy and precision.

Certifications: Gilson maximum permissible errors are guaranteed only when PIPETMAN® pipettors are used with the recommended PIPETMAN® Diamond Tips.




 

ORDER

Expand All / Collapse All
Description Volume Supplier No. VWR Catalog Number Unit Price Quantity
Model F2 2 µL F123770 76178-514 Each Retrieving
Product available on GSA Advantage®
Model F5 5 µL F123771 76178-512 Each Retrieving
Product available on GSA Advantage®
Model F10 10 µL F123772 76178-516 Each Retrieving
Product available on GSA Advantage®
Model F20 20 µL F123604 76178-434 Each Retrieving
Product available on GSA Advantage®
Model F25 25 µL F123775 76178-518 Each Retrieving
Product available on GSA Advantage®
Model F50 50 µL F123778 76178-520 Each Retrieving
Product available on GSA Advantage®
Model F100 100 µL F123784 76178-522 Each Retrieving
Product available on GSA Advantage®
Model F200 200 µL F123605 76178-436 Each Retrieving
Product available on GSA Advantage®
Model F250 250 µL F123787 76178-524 Each Retrieving
Product available on GSA Advantage®
Model F300 300 µL F123788 76178-526 Each Retrieving
Product available on GSA Advantage®
Model F400 400 µL F123789 76178-528 Each Retrieving
Product available on GSA Advantage®
Model F500 500 µL F123790 76178-530 Each Retrieving
Product available on GSA Advantage®
Model F1000 1000 µL F123606 76178-510 Each Retrieving
Product available on GSA Advantage®
Model F5000 5000 µL F123607 76178-532 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Carousel Pipettor Stand (Holds 7) F161401 76178-564 Each Retrieving
Comfort Handle (Blue 3 pack) F161486 76178-506 Pack of 3 Retrieving
Comfort Handle Label (120 labels) F161499 76178-508 Pack of 120 Retrieving
Jimmy Microtube Opener F144983 76178-572 Each Retrieving
PIPETMAN® Coloris Clips (Blue bag of 10) F161305 76178-568 Pack of 10 Retrieving
PIPETMAN® Coloris Clips (Green bag of 10) F161304 76178-566 Pack of 10 Retrieving
PIPETMAN® Coloris Clips (Mixed Color bag of 10) F161301 76178-118 Pack of 10 Retrieving
PIPETMAN® Coloris Clips (Red bag of 10) F161302 76178-120 Pack of 10 Retrieving
PIPETMAN® Coloris Clips (White bag of 10) F161306 76178-570 Pack of 10 Retrieving
PIPETMAN® Coloris Clips (Yellow bag of 10) F161303 76178-122 Pack of 10 Retrieving
Reagent Reservoir 25 mL (10 packs of 10) F267660 76178-114 Pack of 10 Retrieving
Reagent Reservoir 50 mL (10 packs of 10) F267670 76178-116 Pack of 10 Retrieving
Single Pipette Holder F161406 76178-110 Each Retrieving
Trio Pipettor Stand (Holds 3) F161405 76178-108 Each Retrieving
Universal Multichannel Stand F161417 76178-112 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty