Xanthine monosodium salt monohydrate 96+% (dry basis)
Supplier: Thermo Fisher Scientific
Synonyms:
2,6-Dihydroxypurine sodium salt monohydrate
J23465-22
J23465-09
J23465-06
101171-238EA
75.92
USD
101171-238
101173-138
101175-220
Xanthine monosodium salt monohydrate 96+% (dry basis)
Xanthine monosodium salt monohydrate
Formula:
C₅H₃N₄O₂Na·H₂O MW: 192.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069669 CAS Number: 1196-43-6 |
Specification Test Results
Assay (UV/Vis) | ≥96.0% (Dry Basis) |
Reference Constants Epsilonmax (pH 10, NRC) | 9.3×10³ |
Reference Constants Epsilon₂₆₀ (pH 10, DBR) | 5.2×10³ |
Reference Constants Lambdamax (pH 10, NRC) | 277 nm |
Identification (FTIR) | Conforms |
Water Content (Karl Fischer Titration) | ≤12.0% |
Spectral Ratio (pH 10.0) A₂₅₀/A₂₆₀ | 1.29 ±0.06 |
Spectral Ratio (pH 10.0) A₂₈₀/A₂₆₀ | 1.71 ±0.05 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...