Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate 90+% (by HPLC)

J23160-MC J23160-09
103728-118EA 1732.63 USD
103728-118 101170-196
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate 90+% (by HPLC)
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Formula: C₉H₁₂N₂Na₃O₁₅P₃·2H₂O
MW: 586.12
Storage Temperature: Freezer
CAS Number: 116295-90-0
EINECS: 243-347-5

Order Now

Specification Test Results

Form White crystalline powder
Assay (HPLC) ≥ 90.0%
Assay (UV/Vis) 100% ± 3% (Dry Basis)
Reference Constants Epsilonmax (pH 7, NRC) 10.0 x 10³
Reference Constants Epsilon₂₆₀ (pH 7, DBR) 9.9 x 10³
Reference Constants Lambdamax (pH 7, NRC) 262 nm
Residual Water ≤ 15.0%
Lambda-max (UV-Vis) Lambdamax (pH 7.0) 262 nm ± 1 nm
Molar Extinction Coefficient (Molar Absorptivity) Epsilonmax (pH 7.0) [10.0 ± 0.3] x 10³
Spectral Ratio (pH 7.0) A₂₅₀/A₂₆₀ 0.60 ± 0.90
Spectral Ratio (pH 7.0) A₂₈₀/A₂₆₀ 0.30 ± 0.45

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR