D-(+)-Galactose ≥99%
Supplier: Thermo Fisher Scientific
Synonyms:
(2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal
, (2R,3S,4S,5R)-Pentahydroxyhexanal
J15995-A7
J15995-A1
101170-884EA
386.31
USD
101170-884
101171-630
D-(+)-Galactose ≥99%
D(+)-Galactose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 164…168 °C Density: 1.50 Storage Temperature: Ambient |
MDL Number:
MFCD00151230 CAS Number: 59-23-4 EINECS: 200-416-4 Merck Index: 14,04335 |
Specification Test Results
Assay (HPLC) | ≥99.0% |
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.5% |
Loss on Drying | ≤0.5% |
Optical Rotation (c=4 in H₂O and NH₃) | ≥77° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...