4-(Dimethylamino)cinnamaldehyde 98+%, yellow crystalline powder

Warning

Synonyms: p-Dimethylamino-Cinnamaldehyde

J14872-14 J14872-22
101172-452EA 197.9 USD
101172-452 103728-090
4-(Dimethylamino)cinnamaldehyde 98+%, yellow crystalline powder
4-(Dimethylamino)cinnamaldehyde
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Melting Pt: 137…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00007002
CAS Number: 6203-18-5
EINECS: 228-267-0

Order Now

Specification Test Results

Form Bright yellow/gold crytsalline powder
Assay (unspecified) ≥ 98.0%
Identification (FTIR) Conforms
Melting Point 137° - 142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR